missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Drum-Containment Pallets and Platforms

Grating removes easily for cleaning
Supplier: Eagle™ 1647
Description
Pallets and platforms contain spills and overflows of hazardous materials, keeping floors contamination-free.
- Solid polyethylene construction provides excellent chemical resistance
- Consist of translucent tub in safety yellow, and black grating with 1.75 in. (4.4 cm) square openings
- Pallet units only are forkliftable
- Poly ramp can be used with 4-drum inline platform, and with 6-drum and 8-drum low-profile platforms
Compliance
Meet U.S. EPA requirements for Secondary Spill Containment of Hazardous Materials 40CFR264.175.
Chemical Identifiers
| Yellow | |
| HDPE | |
| Contain spills and overflows of hazardous materials, keeping floors contamination |
Specifications
| EPA, 40 CFR 264.175 | |
| 63 gal. | |
| HDPE | |
| Contain spills and overflows of hazardous materials, keeping floors contamination | |
| 3629 kg | |
| 263 cm | |
| 17 cm | |
| 67 cm | |
| 29 kg |
| Yellow | |
| 238 L | |
| 4 Drum Pallet, without Drain | |
| 8000 lb. | |
| 103.5 in. | |
| 6.5 in. | |
| 26.25 in. | |
| 64 lb. |