Learn More
Eagle™ Drum-Containment Pallets and Platforms
GREENER_CHOICE
Grating removes easily for cleaning
Supplier:  Eagle™ 1689
 View more versions of this product
Catalog No. 189994523
Only null left
Description
Description
Pallets and platforms contain spills and overflows of hazardous materials, keeping floors contamination-free.
  • Solid polyethylene construction provides excellent chemical resistance
  • Consist of translucent tub in safety yellow, and black grating with 1.75 in. (4.4 cm) square openings
  • Pallet units only are forkliftable
  • Poly ramp can be used with 4-drum inline platform, and with 6-drum and 8-drum low-profile platforms
Compliance
Meet U.S. EPA requirements for Secondary Spill Containment of Hazardous Materials 40CFR264.175.
Specifications
Chemical Identifiers
Yellow
HDPE
Contain spills and overflows of hazardous materials, keeping floors contamination
Specifications
EPA, 40 CFR 264.175
HDPE
Contain spills and overflows of hazardous materials, keeping floors contamination
1500 lb.
45.5 x 32 x 8 in. (116 x 81 x 20.32 cm)
116 cm
81 cm
20.32 cm
15 kg
Yellow
Poly Pallet Ramp
Eagle 4 drum inline platform, and with 6 drum and 8 drum low profile platforms
680 kg
45.5 in.
32 in.
8 in.
32 lb.
Product Suggestions
Product Suggestions