missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Drum-Containment Pallets and Platforms

Grating removes easily for cleaning

Supplier:  Eagle™ 1686

 View more versions of this product

Catalog No. 18886A


Only null left

Description

Description

Pallets and platforms contain spills and overflows of hazardous materials, keeping floors contamination-free.

  • Solid polyethylene construction provides excellent chemical resistance
  • Consist of translucent tub in safety yellow, and black grating with 1.75 in. (4.4 cm) square openings
  • Pallet units only are forkliftable
  • Poly ramp can be used with 4-drum inline platform, and with 6-drum and 8-drum low-profile platforms

Compliance

Meet U.S. EPA requirements for Secondary Spill Containment of Hazardous Materials 40CFR264.175.

Specifications

Chemical Identifiers

Yellow
HDPE
Contain spills and overflows of hazardous materials, keeping floors contamination

Specifications

EPA, 40 CFR 264.175
85 gal.
HDPE
Contain spills and overflows of hazardous materials, keeping floors contamination
3629 kg
199 cm
17 cm
131 cm
Yellow
322 L
6 Drum Pallet, without Drain
8000 lb.
78.25 in.
6.5 in.
51.5 in.
85 gal.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions