missing translation for 'onlineSavingsMsg'
Learn More

(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane 95.0+%, TCI America™

Catalog No. T29771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29771G 1 g
T29775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29771G Supplier TCI America Supplier No. T29771G

Chemical Identifiers

CAS 55364-28-8
Molecular Formula C6H11F3Si
Molecular Weight (g/mol) 168.234
InChI Key MBCUJGNIYCDYKD-UHFFFAOYSA-N
Synonym Ikeda - Omote Reagent
PubChem CID 71441560
IUPAC Name trimethyl(3,3,3-trifluoroprop-1-enyl)silane
SMILES C[Si](C)(C)C=CC(F)(F)F

Specifications

Color Colorless
Boiling Point 69°C
Quantity 1 g
UN Number 1993
Formula Weight 168.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane