missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1%

Catalog No. 361056784
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361056784 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361056784 Supplier Invitrogen™ Supplier No. G8321ST

Provides everything needed to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.

The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%, provides everything you need to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.

The kit includes:

  • 10 E-Gel agarose gels with SYBR Safe, 1% (Cat. No. A42100)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)

Specifications

Content And Storage • 1 electrophoresis device
• 1 camera device
• 10 E-Gel agarose gels with SYBR Safe, 1%
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses

Store at room temperature.

Type Electrophoresis System Starter Kit
Quantity 1 Kit
Shipping Condition Room Temperature
Max. Voltage 48 VDC
Voltage 240 VAC

For Research Use Only. Not for use in diagnostic procedures.