Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, EX DC 1% DFS Item
The Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, EX DC 1%, provides everything you need to quickly start using precast E-Gel™ EX Double Comb 1% agarose gels on the system for a streamlined gel electrophoresis and gel imaging workflow.
Supplier:  Invitrogen™ G8331ST
Catalog No. G8331ST
Only null left
Explore available promotions
Description
Description
The kit includes:
  • Ten E-Gel™ EX Double Comb Agarose Gels, 1% (Cat. No. A42345)
  • E-Gel™ Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel™ 1-Kb Plus Express DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 10488091)
Specifications
Specifications
• Ten E-Gel EX Double Comb Agarose Gels, 1% (Cat. No. A42345)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
Store at room temperature.
Electrophoresis System Starter Kit
1 kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.