missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 2% DFS Item

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 2%, provides everything you need to quickly start using precast 1% E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the system for a streamlined gel electrophoresis and gel imaging workflow.

Supplier:  Invitrogen™ G8372ST

Catalog No. G8372ST


Only null left
Explore available promotions

Description

Description

The kit includes:

  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
  • E-Gel™ Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel™ Low Range Quantitative DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 12373031)
Specifications

Specifications

• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel Low Range Quantitative DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 12373031)

Store at room temperature.
Mini
Electrophoresis System Starter Kit
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.