Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 1%
Catalog No. G8371ST
Encompass
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
G8371ST 1 kit
1 options
Catalog No. G8371ST Supplier Invitrogen™ Supplier No. G8371ST
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 1%, provides everything you need to quickly start using precast 1% E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
  • E-Gel™ Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel™ 1-Kb Plus Express DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 10488091)
Specifications
Content And Storage • Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
Store at room temperature.
Gel Size Mini
Type Electrophoresis System Starter Kit
Quantity 1 kit
For Research Use Only. Not for use in diagnostic procedures.