Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 2%
The Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 2%, provides everything you need to quickly start using precast E-Gel™ EX Double Comb 2% agarose gels on the device.
Supplier: Invitrogen™ G8132ST
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX DC 2%, provides everything you need to quickly start using precast E-Gel EX Double Comb 2% agarose gels on the device.
The kit includes:
- Ten E-Gel EX Double Comb Agarose Gels, 2% (Cat. No. A42346)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
Specifications
| • Ten E-Gel EX Double Comb Agarose Gels, 2% (Cat. No. A42346) • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100) • E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099) Store at room temperature. |
|
| Electrophoresis System Starter Kit | |
| 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.