missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX 1%
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel EX 1% agarose gels.
Supplier: Invitrogen™ G8141ST
Includes: E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX 1%, provides everything you need to quickly start using precast E-Gel EX 1% agarose gels on the device.
The kit includes:
- 10 E-Gel EX agarose gels, 1% (Cat. No. G401001)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 1 Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
- Gel opening knife (Cat. No. EI9010)
Warranty
1 year warranty
Specifications
| • 1 electrophoresis device • 10 E-Gel EX 1% gels • 1 vial E-Gel 1 Kb Plus Express DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 knife Store at room temperature. |
|
| 1 Kit | |
| 48 VDC |
| Electrophoresis System Starter Kit | |
| Room Temperature | |
| 240 VAC |
For Research Use Only. Not for use in diagnostic procedures.