missing translation for 'onlineSavingsMsg'
Learn More

(E)-5-(2-Bromovinyl)-2'-deoxyuridine 98.0+%, TCI America™

Catalog No. B3404100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3404100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3404100MG Supplier TCI America Supplier No. B3404100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69304-47-8
Molecular Formula C11H13BrN2O5
Molecular Weight (g/mol) 333.14
MDL Number MFCD00058585
InChI Key ODZBBRURCPAEIQ-PIXDULNESA-N
Synonym brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
PubChem CID 446727
IUPAC Name 5-[(1E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 100 mg
Formula Weight 333.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (E)-5-(2-Bromovinyl)-2′-deoxyuridine
RTECSNumber YU7355000
TSCA No
Recommended Storage Refrigerator