missing translation for 'onlineSavingsMsg'
Learn More

(E)-3,4-Dihydroxybenzylideneacetone, 97%

Catalog No. p-7028533
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2590103 1 g
AAH2590106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2590103 Supplier Thermo Scientific Chemicals Supplier No. H2590103
Only null left

CAS: 123694-03-1 | C10H10O3 | 178.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123694-03-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00916691
InChI Key YIFZKRGUGKLILR-UHFFFAOYSA-N
Synonym 3-buten-2-one,4-3,4-dihydroxyphenyl-, 3e, 3,4-dihydroxybenzalacetone, acmc-20dpit
PubChem CID 53446072
IUPAC Name 4-(3,4-dihydroxyphenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Melting Point 173°C to 175°C
Quantity 1 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material (E)-3,4-Dihydroxybenzylideneacetone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only