Learn More
Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta)
Form: Approx. 0.27g/mm, Cast rod, diameter is approximate | CAS: 7429-91-6 | Dy | 162.50 g/mol
Supplier: Thermo Scientific Chemicals 040285DH
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dysprosium | |
| 7429-91-6 | |
| 25 mm | |
| Dy | |
| ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
| [Dy] | |
| 162.50 | |
| CHEBI:33377 | |
| Gray to silver rod |
| Gray to Silver | |
| Cast Rod | |
| 99.7% (Metals basis excluding Ta) | |
| MFCD00010982 | |
| KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
| dysprosium | |
| 23912 | |
| Odorless |
Chemical Identifiers
| 7429-91-6 | |
| 162.50 | |
| KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
| 23912 | |
| dysprosium |
| Dy | |
| MFCD00010982 | |
| ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
| CHEBI:33377 | |
| [Dy] |
Safety and Handling
EINECSNumber : 231-073-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only