missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium pieces, 99.9% (REO)

Catalog No. p-7048191
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0010609 10 g
AA0010618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0010609 Supplier Thermo Scientific Chemicals Supplier No. 00010609
Only null left

CAS: 7429-91-6 | Dy | 162.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Silver to Gray
Quantity 10 g
Odor Odorless
Form Pieces
UN Number UN3178
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Dysprosium pieces
Hazard Category Danger
Hazard Statement GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.
Precautionary Statement GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 231-073-9
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only