missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium ingot, 99.8% (REO)

Catalog No. AA0010214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0010214 25 g
AA0010222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0010214 Supplier Thermo Scientific Chemicals Supplier No. 00010214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7429-91-6 | Dy | 162.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Quantity 25 g
Odor Odorless
Packaging Poly bottle
Form Ingot
Percent Purity 99.8%
Assay (REO)
Chemical Name or Material Dysprosium ingot
EINECSNumber 231-073-9
TSCA Yes
storageNote1
  • Packaged in air stable poly bottle
Recommended Storage Ambient temperatures

RUO – Research Use Only