missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium(III) isopropoxide, 99.9% (REO), 5% w/v in toluene/isopropanol
Description
Dysprosium(III) isopropoxide is used as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Linear Formula | Dy[OCH(CH3)2]3 |
| Quantity | 25 mL |
| UN Number | UN1993 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 339.76 |
| Percent Purity | 99.9% |
| Concentration or Composition (by Analyte or Components) | 5% w/v in toluene/isopropanol |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | H225-H304-H315-H318-H335-H336-H361d-H373 |
| Hazard Statement | GHS H Statement H225-H304-H361-H373-H315-H319-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only