missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) isopropoxide, 99.9% (REO), 5% w/v in toluene/isopropanol

Catalog No. AA36585AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36585AE 100 mL
AA36585AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36585AE Supplier Thermo Scientific Chemicals Supplier No. 036585AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6742-68-3 | C9H24DyO3 | 342.788 g/mol

Dysprosium(III) isopropoxide is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6742-68-3
Molecular Formula C9H24DyO3
Molecular Weight (g/mol) 342.788
MDL Number MFCD00049784
InChI Key IWDABVYEISORRX-UHFFFAOYSA-N
Synonym dysprosium iii i-propoxide-dy reo
PubChem CID 131675877
IUPAC Name dysprosium;propan-2-ol
SMILES CC(C)O.CC(C)O.CC(C)O.[Dy]

Specifications

Color Brown
Linear Formula Dy[OCH(CH3)2]3
Quantity 100 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 339.76
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Dysprosium(III) isopropoxide
Show More Show Less
Hazard Category H225-H304-H315-H318-H335-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only