missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) acetate tetrahydrate, REacton™, 99.99% (REO)

Catalog No. AA1131409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1131409 10 g
AA1131418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1131409 Supplier Thermo Scientific Chemicals Supplier No. 01131409

CAS: 15280-55-4 | C6H17DyO10 | 411.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15280-55-4
Molecular Formula C6H17DyO10
Molecular Weight (g/mol) 411.69
MDL Number MFCD00013033
InChI Key SIYZPSYZNCQFDV-UHFFFAOYSA-K
IUPAC Name dysprosium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Dy+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point 120°C (decomposition)
Physical Form Crystalline
Quantity 10 g
Formula Weight 411.68 (339.62 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) acetate tetrahydrate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only