missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium foil, 0.25mm (0.01 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

$949.74 - $949.74

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA12381FI
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
012381FI
50 x 50 mm
Each for $949.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7429-91-6
162.50
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
CHEBI:33377
[Dy]

Specifications

7429-91-6
Odorless
Foil
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
[Dy]
162.50
CHEBI:33377
Dysprosium foil, 0.25mm (0.01 in.) thick
50 x 50 mm
≈1.33g/25x25mm
Dy
UN3178
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
99.9%
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-073-9

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only