missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium foil, 0.1mm (0.004in) thick, 99.9% (REO)

Catalog No. p-4824706
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12382FF 25 x 25 mm
AA12382FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12382FF Supplier Thermo Scientific Chemicals Supplier No. 012382FF
Only null left

Form: Approx. 0.53g/25 x 25mm | CAS: 7429-91-6 | Dy | 162.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Gray to Silver
Physical Form Foil
Quantity 25 x 25 mm
Assay Percent Range 99.90%
Odor Odorless
Appearance Gray to silver foil
Chemical Name or Material Dysprosium
Hazard Category H260-H228; In contact with water releases flammable gases which may ignite spontaneously. Flammable solid.
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-073-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only