missing translation for 'onlineSavingsMsg'
Learn More

Dyphylline

Catalog No. AAJ6316722
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6316722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6316722 Supplier Thermo Scientific Chemicals Supplier No. J6316722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator | CAS: 479-18-5 | C10H14N4O4 | 254.246 g/mol

An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 479-18-5
Melting Point 160°C to 165°C
Color White
Molecular Formula C10H14N4O4
Quantity 100 g
MDL Number MFCD00005759
Beilstein 284563
Merck Index 14,3479
Synonym dyphylline, diprophylline, diphylline, 7-2,3-dihydroxypropyl theophylline, diprophyllin, glyphylline, neothylline, lufyllin, aristophyllin, diprofilline
Solubility Information Soluble in water
InChI Key KSCFJBIXMNOVSH-UHFFFAOYSA-N
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
IUPAC Name 7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
Molecular Weight (g/mol) 254.246
PubChem CID 3182
ChEBI CHEBI:4728
Formula Weight 254.24
Physical Form Powder
Chemical Name or Material Dyphylline
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 207-526-1
RTECSNumber XH5100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only