missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Dynamics System Starter Set for Physics and Mechanics

Catalog No. S27569
Change view
Click to view available options
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S27569 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27569 Supplier Eisco™ Supplier No. DYSYS

This Dynamics System Starter Set is an economical way to teach dynamics principles such as position, velocity, acceleration, force, and elastic. In elastic collisions, it is suitable for teaching momentum, conservation of energy, and much more.

  • Kit contains everything needed to start to teach dynamics
  • Shipped in 2 boxes due to size

Specifications

Product Type Dynamics System Starter Set
Description Shipped in 2 Boxes
For Use With (Application) Teaching Physics and Mechanics Principles
Includes (1) 1.2 m Track with Pulley, (1) Standard Trolley, (1) Spring Loaded Trolley, (2) Masses, (10) 10 g Masses, (1) Ticker Tape Timer, (5) Spare Tapes, (1 Pk.) Carbon Discs, (1 Reel) Cotton, (1) Spirit-Filled Level, and (1) Stopwatch
Quantity 1 Kit