Learn More
Dynamic Diagnostics Safety Shields
Catalog No. 22899025
Encompass
Change view
Click to view available options
Product Type:
Clamp-on Safety Shield
Magnetic Safety Shield
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22899025 Clamp-on Safety Shield
22899024 Magnetic Safety Shield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22899025 Supplier Dynamic Diagnostics Supplier No. 300018
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Magnetic or clamp-on splash shields with gooseneck arm for exact angle positioning
  • Exact angle positioning
  • 12 x 8.5 in.
  • Clear polycarbonate
  • UV protection from 200-360 nanometers
  • Available as magnetic base or a rubber-coated, clamp-on base
Specifications
Product Type Clamp-on Safety Shield
No. per Pack 1 Ea.