missing translation for 'onlineSavingsMsg'
Learn More

Dynalon™ Non-Hazardous-Waste Lab Disposal Box

Catalog No. 1417420
Change view
Click to view available options
Length (English):
12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
1417420 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1417420 Supplier Dynalon™ Supplier No. 797055

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy identification

Non-Hazardous Disposal Box features colorful graphics for quick identification.

  • Assembly instructions on each box
  • Tough 2 mil polyethylene liner
  • Folding design for shipping and storage
  • Closure assures a safe, hygienic disposal
  • Supplied in a case of 6 boxes

Specifications

Capacity (Metric) 60L
Closure Type Lid
Description Non-Hazardous Disposal Box
Diameter (Metric) Outer 177.8 mm
Diameter (English) Outer 7 in.
Capacity (English) 16 gal.
Color White, Green
Dimensions (L x W x H) 12 x 12 x 27 in.
Height (Metric) 68.58 cm
Length (Metric) 30.48 cm
Quantity 6/Cs.
Shape Rectangle
Width (Metric) 30.48 cm
Autoclavable Not Autoclavable
Includes Polyethylene Liner
Liner Material Polyethylene
For Use With (Application) For nonhazarous lab waste
Width (English) 12in
Length (English) 12 in.
Height (English) 27in
Material Corrugated Cardboard
Product Type Waste Lab Box
Show More Show Less