Learn More
Invitrogen™ Dynabeads™ Untouched™ Human CD4 T Cells Kit
Use this kit to isolate pure and viable untouched CD4+ T-cells from PBMC by negative isolation. The kit depletes CD8+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes.
$572.00 - $1,776.00
Specifications
| Cell Type | T Cells (CD4) |
|---|---|
| Product Type | Cell Isolation Kit |
| Final Product Type | Cells |
| Isolation Technology | Negative Isolation |
| Product Line | DYNAL, Dynabeads, Untouched |
Description
The kit depletes CD8+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes. The negatively isolated human CD4+ T-cells are left in sample and have not been in contact with Dynabeads. The bead and antibody-free CD4+ T-cells are truly untouched, and in perfect shape for any functional assay and application.
- High purity, recovery and viability of human untouched CD4+ T cells
- Easy to use and to scale up
- No columns required
- Not high throughput compatible
A negative isolation strategy isolates pure and truly untouched cells. When maximum cell purity is critical, a positive cell isolation strategy is recommended.
Specifications
| T Cells (CD4) | |
| Cells | |
| DYNAL, Dynabeads, Untouched | |
| Room Temperature |
| Cell Isolation Kit | |
| Negative Isolation | |
| Research Grade | |
| Human |
For Research Use Only. Not for use in diagnostic procedures.