Learn More
Invitrogen™ Dynabeads™ CD4
Dynabeads CD4 are superparamagnetic beads coupled with an anti-human CD4 antibody that enable easy isolation or depletion of human CD4+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, PBMC, and tissue digests.
Supplier: Invitrogen™ 11146D
Description
Dynabeads CD4 are superparamagnetic beads coupled with an anti-human CD4 antibody that enable easy isolation or depletion of human CD4+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, peripheral blood mononuclear cells (PBMCs), and tissue digests. Advantages of Dynabeads CD4:
- Fast and efficient isolation of CD4+ T cells from any sample—no columns required
- High purity, yield, and viability of positively isolated CD4+ T cells for molecular downstream assays
- Fast and efficient depletion of CD4+ T cells from any sample
About Dynabeads CD4
Dynabeads CD4 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD4 membrane antigen mainly expressed on human T helper cells. Because of the bead size, Dynabeads CD4 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads CD4 will also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD4+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human CD4 product offerings.
Magnetic bead to based separation offers easy handling
Dynabeads CD4 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.
Automation-ready for KingFisher purification systems
The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.
Learn more about Dynabeads products
- Find Dynabeads products for a whole range of applications.
- Find magnets for Dynabeads separations.
Specifications
| T Cells | |
| Brown | |
| Magnetic Polystyrene | |
| Processes ∼8x 10^8 cells total | |
| Dynabeads | |
| 2 mL | |
| Room Temperature | |
| Human |
| This Product Contains: 2 mL Dynabeads™ CD4 coated with anti-CD4 monoclonal antibody. Storage Conditions: 2°C to 8°C. Do not freeze. |
|
| Cell Depletion, Positive Isolation for Molecular Applications | |
| Yes | |
| >95% | |
| Research Grade | |
| PBMC, Blood | |
| 1 x 10^7 PBMCs per isolation | |
| 4.5 μm |
For Research Use Only. Not for use in diagnostic procedures.