GA International DYMO-Compatible Paper Labels 0.5"" / 12.7mm Circle Blue Ambient temperature
Catalog No. 5000570861
DYMO compatible paper labels for general use applications and laboratory organization. Ideal for office use on file folders, boxes, and shipping containers. Not suitable for deep-freeze applications.
Catalog No.
5000570861
Supplier
GA InternationalSupplier No.
ECY050BL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
DYMO compatible paper labels for general use applications and laboratory organization. Ideal for office use on file folders, boxes, and shipping containers. Not suitable for deep-freeze applications.