missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dyclonine Hydrochloride 98.0+%, TCI America™
$143.54 - $419.41
Chemical Identifiers
| CAS | 536-43-6 |
|---|---|
| Molecular Formula | C18H28ClNO2 |
| Molecular Weight (g/mol) | 325.88 |
| MDL Number | MFCD00035386 |
| InChI Key | KNZADIMHVBBPOA-UHFFFAOYSA-N |
| Synonym | dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride |
| PubChem CID | 68304 |
| ChEBI | CHEBI:4725 |
| IUPAC Name | hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride |
| SMILES | [H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1 |
Chemical Identifiers
| 536-43-6 | |
| 325.88 | |
| KNZADIMHVBBPOA-UHFFFAOYSA-N | |
| 68304 | |
| hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride |
| C18H28ClNO2 | |
| MFCD00035386 | |
| dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride | |
| CHEBI:4725 | |
| [H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1 |
Specifications
| 536-43-6 | |
| White | |
| MFCD00035386 | |
| dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride | |
| [H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1 | |
| 325.88 | |
| CHEBI:4725 | |
| ≥98.0% (HPLC,T) | |
| Dyclonine Hydrochloride |
| 173°C | |
| C18H28ClNO2 | |
| 5 g | |
| KNZADIMHVBBPOA-UHFFFAOYSA-N | |
| hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride | |
| 68304 | |
| 325.88 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UG8750000
TSCA : No
Recommended Storage : Refrigerator