missing translation for 'onlineSavingsMsg'
Learn More

Dyclonine Hydrochloride 98.0+%, TCI America™

Catalog No. D43035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43035G 5 g
D430325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43035G Supplier TCI America Supplier No. D43035G

Chemical Identifiers

CAS 536-43-6
Molecular Formula C18H28ClNO2
Molecular Weight (g/mol) 325.88
MDL Number MFCD00035386
InChI Key KNZADIMHVBBPOA-UHFFFAOYSA-N
Synonym dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride
PubChem CID 68304
ChEBI CHEBI:4725
IUPAC Name hydrogen 1-(4-butoxyphenyl)-3-(piperidin-1-yl)propan-1-one chloride
SMILES [H+].[Cl-].CCCCOC1=CC=C(C=C1)C(=O)CCN1CCCCC1

Specifications

Melting Point 173°C
Color White
Quantity 5 g
Formula Weight 325.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dyclonine Hydrochloride
RTECSNumber UG8750000
TSCA No
Recommended Storage Refrigerator