missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dyclonine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 173°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 325.88 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dyclonine Hydrochloride |
Safety and Handling
| RTECSNumber | UG8750000 |
| TSCA | No |
| Recommended Storage | Refrigerator |