Learn More
DWK Life Sciences DURAN™ Grey Bromobutyl Rubber Stopper, Straight Plug, for GL 25 or GL 45 Laboratory Bottles
Catalog No. 09841325
Change view
Click to view available options
Standard Taper Joint:
GL 25
GL 45
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
09841325 GL 25
09841311 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09841325 Supplier DWK Life Sciences Supplier No. 292061301
Only null left
DURAN™ bromobutyl rubber closure provide a gas tight seal for all GL 25 and GL 45 laboratory bottles.
DURAN™ bromobutyl rubber closure provide a gas tight seal for all GL 25 and GL 45 laboratory bottles. Bromobutyl rubber is essentially impermeable to most gases, and provides a controlled environment inside the glass bottle for oxygen sensitive materials. Useful for maintaining anaerobic culture conditions. Butyl rubber allows for multiple punctures providing easy access to the contents with a syringe.
  • Bromobutyl rubber elastomer (USP <381> / EP 3.2.9 Type I closure for chemical and penetration tests)
  • Gas tight seal provides controlled environment in the bottle
  • Certificate of Conformance (CoC)
  • Multiple punctures providing easy access to the contents with a syringe
  • Elastic material for effective sealing and reliable self-sealing after needle piercing
  • Material is virtually free of organics that might leach into the incubation medium
  • Stopper shape and material properties optimised to fit DURAN™ GL 25 and GL 45 bottles
  • Wide usable temperature range: -50 to +121°C
  • Suitable for autoclaving / steam sterilising at 121°C for 20 minutes
Specifications
Autoclavable Autoclavable
Shape Cylindrical
For Use With (Application) Culture of anaerobic microorganisms; Storage of anaerobic culture media; Aerobic methane oxidation (MOx) research; Storage of oxygen or moisture sensitive materials such as APIs; Nitrogen Blanketing; Applications requiring gas purging or sample transfer via a syringe
Color Gray
Material BIIR
Quantity 10/Pk.
Standard Taper Joint GL 25
Diameter (Metric) 22.5 mm