missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Screw Caps for GL 45 Laboratory Glass Bottles
Description
- Typical applications: injection or removal of media.
Specifications
Specifications
| Color | Red |
| Material | PBT |
| Thread Size (Metric) | GL 45 |
| Neck Finish | 9 mm |
| For Use With (Equipment) | GL45 laboratory glass bottles |
| Product Type | Screw Cap with Aperture |