Learn More
DWK Life Sciences™ DURAN™ protect+ Graduated Clear Laboratory Bottle with Wide Mouth
Multi-functional glass wide-mouth laboratory bottles with protective safety coating.
Supplier: DWK Life Sciences™ 218654484
Description
The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.
- Manufactured from Polyamide-coated DURAN™ borosilicate glass 3.3
- DURAN™ is a neutral glass, classified as a Glass Type I according to USP, EP (3.2.1) and JP, and meets ASTM E438, Type I, Class A requirements
- Reduce the hazards of broken glass and liquid splashes in the event of breakage
- The external polymer coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL and less)
- Polyamide 12 safety coating has a high chemical resistance
- Protective coating provides an enhanced safer grip
- Coating is Cleanroom certified Fraunhofer IPA TESTED DEVICE™
- Certified FDA and EU food contact Safe coating material
- Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
- Supplied with push-on dust cover
Specifications
| 500 mL | |
| Clear | |
| Borosilicate Glass (3.3) | |
| Non-sterile | |
| GLS 80 | |
| No | |
| Wide | |
| 148 mm | |
| 80 | |
| Not Autoclavable |
| 16.9 oz. | |
| Preparation of media and reagents, safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs), sample or harvest collection container, culturing vessel, storage of media and reagents, transport and shipping, Liquid handling reservoir | |
| 10 Bottles | |
| 0.52 kg | |
| CoC | |
| Yes | |
| Round | |
| Laboratory Bottle | |
| 101 mm |