Learn More
DWK Life Sciences™ DURAN™ protect+ Graduated Amber Laboratory Bottle with Wide Mouth
Multi-functional glass wide-mouth laboratory bottles with protective safety coating.
$1586.71 - $4018.12
Specifications
| Color | Amber |
|---|---|
| For Use With (Application) | Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping |
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Thread Size (Metric) | GLS 80 |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
13-299-158
|
DWK Life Sciences™
218663686 |
250 mL |
Case of 10 Each for $3,315.44
|
|
|||||
|
13-299-157
|
DWK Life Sciences™
218664485 |
500 mL |
Case of 10 Each for $3,371.97
|
|
|||||
|
13-299-156
|
DWK Life Sciences™
218665481 |
1 L |
Case of 10 Each for $4,018.12
|
|
|||||
|
13-299-154
|
DWK Life Sciences™
218666983 |
3.5 L |
Case for $1,586.71
|
|
|||||
|
13-299-153
|
DWK Life Sciences™
218667388 |
5 L |
Case for $2,272.59
|
|
|||||
Description
The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.
- Manufactured from ambered borosilicate glass 3.3 and polyamide
- Compatible with matching cap system for particularly tight sealing and simple, clean pouring out
- protect+ safety coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
- UV protection USP (Spectral Transmission) and EP (3.2.1) compliant
- Less than 10% spectral transmission between 290 nm and 450 nm
- The only external application of the amber colour and safety coating ensures that internal surfaces have standard DURAN™ glass properties
- Supplied as safety coated glass bottle only (screw caps and pouring rings must be ordered separately)
- Easy-to-read scale with graduation marks to estimate the content
- Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
- Coating process does not use any volatile organic compounds (VOCs) or isocyanates
Specifications
| Amber | |
| Borosilicate Glass (3.3) | |
| GLS 80 | |
| No | |
| Wide | |
| Laboratory Bottle | |
| Not Autoclavable |
| Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping | |
| Non-sterile | |
| CoC | |
| Yes | |
| Round | |
| 80 |