missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT

Catalog No. 09841254
Change view
Click to view available options
Thread Size (Metric):
GL14
GL 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
09841254 GL 25
09841253 GL14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09841254 Supplier DWK Life Sciences Supplier No. 292270902
Only null left

PBT screw cap with an open top and high temperature and chemical resistance.

Open topped cap with central aperture suitable for use with bromobutyl stoppers, silicone piercing septa or glassware connections. More chemically resistant than PP cap.
  • Compatible with GL bottle threads
  • Red PBT offers high temperature and chemical resistance
  • Glass reinforced PBT offers high strength

Specifications

Color Red
Diameter (Metric) Inner Bore Size 15 mm
For Use With (Application) Aseptic reagent delivery and culture sampling using a syringe and needle
Material PBT
Quantity 10/Pk.
Screw Material Polybutylene Terephthalate (PBT) with glass fiber
Thread Size (Metric) GL 25
Product Type Open Top Screw Cap
Closure Material PBT with Glass Fiber
Diameter (Metric) 33 mm
Autoclavable Autoclavable
Show More Show Less