missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GLS 80™ Wide Mouth Bottle Connection System, Screw Cap GLS 80™, 4-Ports, GL 18 Thread

Flexible GLS 80™ connection system for the use of semi-rigid hoses.

$158.75 - $158.75

Specifications

Diameter (Metric) 87 mm
Autoclavable Autoclavable
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Thread Size GL 18
Quantity 1 Ea.
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
09-841-143
Accessories View Documents
DWK Life Sciences
1160169
3.2 mm N/A
Only null left
 
09-841-144
Accessories View Documents
DWK Life Sciences
1160170
6 mm N/A
Only null left
 
09841145
Accessories View Documents
DWK Life Sciences
1160171
8 mm N/A
Only null left
 
09841146
Accessories View Documents
DWK Life Sciences
1160172
10 mm N/A
Only null left
 
Description

Description

Flexible GLS 80™ cap connection system for use with semi-rigid tubing, suitable for DURAN™ GLS 80™ laboratory and media bottles. Five different tube diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation through membrane liter possible. Unused ports can be provided with a blind cap. Exemplary applications: Safe transfer of liquid media within a closed and sterile system.

  • The flexible modular system allows individual combination of the various components according to requirements and application
  • Five different hose diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected
  • NS 29/32 connection in the middle for tight closure
  • Sterile pressure equalisation through membrane filter possible
  • Unused ports can be fitted with a GL 18 blind cap
  • Compatible with all GLS 80 threads and therefore versatile
Specifications

Specifications

87 mm
Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
1 Ea.
Autoclavable
GL 18
Videos
SDS
Documents

Documents