missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles
Description
- Available in blue, green, yellow and grey with match pour rings
Specifications
Specifications
| Color | Yellow |
| Material | Polypropylene |
| Product Type | Screw Cap |
| For Use With (Equipment) | Laboratory glass bottles |
| Closure Size | GL 45 |