missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles

Catalog No. 09841271
Change view
Click to view available options
Color:
Blue
Gray
Green
Red
Yellow
Material:
PBT
Polypropylene
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
09841271 Red PBT
09841270 Blue Polypropylene
09841278 Gray Polypropylene
09841277 Green Polypropylene
09841279 Yellow Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 09841271 Supplier DWK Life Sciences Supplier No. 292402807

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles help simplify media type identification and prevent cross-contamination. Color-coded caps with matching pour rings make it easy to distinguish contents and avoid interchange of screw caps between bottles.

  • GL 45 screw caps designed for DURAN laboratory glass bottles
  • Available in multiple colors including blue, green, yellow, grey and red to aid media type identification
  • Matching colored pour rings included to prevent carry-over of substances between bottles
  • Color-coding simplifies distinguishing between different media types
  • Helps effectively exclude interchanging of screw caps and carry-over of substances

Specifications

Color Red
Material PBT
Sterility Non-sterile
Thread Size (Metric) GL 45
For Use With (Equipment) Laboratory glass bottles
Closure Size GL 45
Autoclavable Autoclavable
Disposable Reusable
Product Type Screw Cap