missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GL 32 Screw Caps for Laboratory Glass Bottles
Description
- Available in blue, green, yellow and grey
Specifications
Specifications
| Color | Red |
| Material | PBT |
| Thread Size (Metric) | GL 32 |
| For Use With (Equipment) | Laboratory glass bottles |
| Product Type | Screw Cap with PTFE-protected Seal |