missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GL 18 pressure equalization set

Catalog No. 02900166
Change view
Click to view available options
Thread Size (Metric):
16 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02900166 16 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02900166 Supplier DWK Life Sciences Supplier No. 1160167

Flexible GLS 80™ connection system for the use of semi-rigid hoses.

Flexible GLS 80™ cap connection system for use with semi-rigid tubing, suitable for DURAN™ GLS 80™ laboratory and media bottles. Five different tube diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation through membrane liter possible. Unused ports can be provided with a blind cap. Exemplary applications: Safe transfer of liquid media within a closed and sterile system.

  • The flexible modular system allows individual combination of the various components according to requirements and application
  • Five different hose diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected
  • NS 29/32 connection in the middle for tight closure
  • Sterile pressure equalisation through membrane filter possible
  • Unused ports can be fitted with a GL 18 blind cap
  • Compatible with all GLS 80 threads and therefore versatile

Specifications

Description Pressure equalisation cap set for 4-port, GL 18
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Material Polypropylene, PTFE
Quantity 1 Ea.
Thread Size (Metric) 16 mm
Neck Finish 16 mm
Diameter (Metric) 87 mm
Autoclavable Autoclavable