missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ Durac™ Specific Gravity / Relative Density (g/cm3) Precision Hydrometers; Traceable to NIST
Measure specific gravity or relative density (g/cm³) of a variety of liquids with these hydrometers. H-B Instrument Durac™ Specific Gravity / Relative Density (g/cm³) Precision Hydrometers are ideal for industries such as food and beverage, chemical, pharmacology, transportation and refrigeration.
Description
- 325mm (12.8 in.) long
- Lead free ballast and glass
- Individually serialized
- Temperature standardization of 60°F (15°C)
- Hand inspected by U.S. quality team
- Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
Chemical Identifiers
| Each | |
| ± 0.001 | |
| Traceable to NIST | |
| Multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST | |
| Lead Free | |
| 15°C |
| Specific Gravity/Relative Density Precision Hydrometer | |
| 0.0005 | |
| 325 mm | |
| Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures | |
| Industries such as food and beverage, chemical, pharmacology, transportation and refrigeration |
Specifications
| Each | |
| ± 0.001 | |
| Traceable to NIST | |
| Multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST | |
| Lead Free | |
| 1.180 to 1.250 |
| Specific Gravity/Relative Density Precision Hydrometer | |
| 0.0005 | |
| 325 mm | |
| Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures | |
| Industries such as food and beverage, chemical, pharmacology, transportation and refrigeration | |
| 15°C |