missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ Durac™ Specific Gravity Precision Hydrometers
Used in food and beverage, chemical, pharmacology, transportation, refrigeration industries
Supplier: H-B Instrument™ B618010800
Alert:
Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Description
- 325mm (12.8 in.) long
- Lead free ballast and glass
- Individually serialized
- Temperature standardization: 60°F (15°C)
- Hand inspected by our U.S. quality team
- Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
Chemical Identifiers
| 10/Pk. | |
| 0.0005 | |
| Lead Free |
| Specific Gravity Precision Hydrometer | |
| 325 mm | |
| 15°C |
Specifications
| 10/Pk. | |
| ±0.001 | |
| 325 mm | |
| 1.060 to 1.130 |
| Specific Gravity Precision Hydrometer | |
| 0.0005 | |
| Lead Free | |
| 15°C |