missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Durac™ Plus™ Total Immersion Thermometer

Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries.

Supplier:  H-B Instrument™ B602051100

Catalog No. S02669


Only null left

Description

Description

  • Certified to NIST standards at specific temperature points
  • Supplied with a Calibration Report stating corrections to one-tenth of the smallest division
  • Feature individual serial numbers
  • Accuracy of 1.5°C or better and blue organic liquid against white back glass
  • Tested and certified in our exclusive triple accredited/registered ISO/IEC 17025:2005
  • A2LA accredited laboratory against equipment whose calibration is traceable to NIST
  • Individually packaged

Specifications

Specifications

ISO/IEC 17025:2005, A2LA, NIST
Total Immersion
305 mm
Blue Organic Liquid
-20°C
±1.5°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
12 in.
Glass
110°C
-20°C to +110°C
1°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions