missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Sleeve

Description
Tyvek sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
- Garments are gamma sterilized to SAL of 10-6
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic at both ends (bicep and wrist) for arm protection
- 18 in. long
- One size fits most
- Full traceability on all sterilized apparel with Certificates of Sterility Available
Specifications
Specifications
| Size | Standard |
| Material | Tyvek |
| Sterility | Sterile |
| Disposable | Single-use |
| Closure Type | Elastic Ends |
| Seam Type | Bound |
| Cuff Style | Elastic |
| Model | IC501B |
| Thickness | 5.0 mil |
| Certifications/Compliance | ASTM F2101 |
| Show More |