missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. 17988048
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
6.4 to 7.2 ft.
Height (Metric):
152.4 to 173.73 cm
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
195.07 to 219.45 cm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
17988048 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
17988043 Small 5 to 5.7 ft. 152.4 to 173.73 cm
17988044 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17988045 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17988046 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
17988047 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
17988049 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
19813221 5X-Large 6.4 to 7.2 ft. 195.07 to 219.45 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17988048 Supplier DuPont™ Supplier No. IC253BWH3X0025CS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Clean-proceesed and sterile coveralls worn in controlled environments protect against particulates and other contaminants.

  • Garments are gamma sterilized to SAL of 10-6.
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing.
  • Full traceability on all sterilized apparel with Certificates of Sterility
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Particle, Non-toxic Particulates
Collar Type Bound
Model IC253BWH
Size (English) Chest 46 to 52 in.
Size (English) Inseam 35 in.
Size (Metric) Chest 116.8 to 132.08 cm
Size (Metric) Inseam 88.9 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids
Length (English) Sleeve 40.75 in.
Length (Metric) Sleeve 103.5 cm
Height (English) 6.2 to 6.6 ft.
Height (Metric) 188.97 to 201.16 cm
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less