missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Clean-proceesed and sterile coveralls worn in controlled environments protect against particulates and other contaminants.
Supplier: DuPont™ IC253BWHXL0025CS
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Clean-proceesed and sterile coveralls worn in controlled environments protect against particulates and other contaminants.
- Garments are gamma sterilized to SAL of 10-6.
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle.
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing.
- Full traceability on all sterilized apparel with Certificates of Sterility
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Covered Elastic | |
| Zipper | |
| Sterile | |
| IC253BWH | |
| 33.5 in. | |
| 85 cm | |
| 5 mil | |
| 38.25 in. | |
| 5.11 to 6.3 ft. |
| X-Large | |
| Tyvek | |
| Covered Elastic | |
| Collared | |
| Bound | |
| Bound | |
| 40 to 43 in. | |
| 101.6 to 109.22 cm | |
| Dolman | |
| Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids | |
| 97.15 cm | |
| 155.75 to 192.04 cm |