missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers
Sterile boot covers create an inherent barrier to dry particles, microorganisms, and nonhazardous liquids.
Supplier: DuPont™ IC458BWHMD0100CS
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Garments are gamma sterilized to SAL of 10-6.
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Pleated rayon outer facing
- 18 in. high boot cover with covered elastic top
- PVC soles provides enhanced skid-resistance and durability
- Full traceability on all sterilized apparel with Certificates of Sterility
- D14238474
- Garments are gamma sterilized to SAL of 10-6.
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Pleated rayon outer facing
- 18 in. high boot cover with covered elastic top
- PVC soles provides enhanced skid-resistance and durability
- Full traceability on all sterilized apparel with Certificates of Sterility
- D15568559
Specifications
| Medium | |
| Yes | |
| Sterile | |
| Anti-skid | |
| 5.4 mil | |
| 30.48 cm | |
| 46.99 cm |
| Tyvek | |
| White | |
| Elastic Top with Ankle Ties | |
| Bound | |
| 12 in. | |
| 18.5 in. |