Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers
Sterile boot covers create an inherent barrier to dry particles, microorganisms, and nonhazardous liquids.
Supplier:  DuPont™ IC457SWHMD01000S
 View more versions of this product
Catalog No. 17-988-093
Only null left
Explore available promotions
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Garments are gamma sterilized to SAL of 10-6.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Pleated rayon outer facing
  • 18 in. high boot cover with covered elastic top
  • PVC soles provides enhanced skid-resistance and durability
  • Full traceability on all sterilized apparel with Certificates of Sterility
  • D14238474
  • Garments are gamma sterilized to SAL of 10-6.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Pleated rayon outer facing
  • 18 in. high boot cover with covered elastic top
  • PVC soles provides enhanced skid-resistance and durability
  • Full traceability on all sterilized apparel with Certificates of Sterility
  • D15568559
Specifications
Specifications
Medium
Yes
Sterile
Anti-skid
5.4 mil
30.48 cm
46.99 cm
Tyvek
White
Elastic Top with Ankle Ties
Serged
12 in.
18.5 in.
Product Suggestions
Product Suggestions