missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 447 Boot Covers

Catalog No. 19039870
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Sterility:
Non-sterile
Sterile
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility
19039870 Medium Non-sterile
19039728D X-Large Non-sterile
19039852 Large Sterile
19039851 Medium Sterile
19039853 X-Large Sterile
19039854 2X-Large Sterile
19039871 Large Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19039870 Supplier DuPont™ Supplier No. IC447SWHMD01000B
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gripper™ soles resists slipping on slick surfaces

  • Antistatic and low linting
  • Provides barrier to dry particulates, nonhazardous liquids and microorganisms
  • Coarsely textured interior to help prevent movement inside
  • PVC Gripper soles provide excellent barrier to liquid penetration
  • 18 in. (45.7cm) high Tyvek™ uppers with elastic leg and ankle openings
  • Serged seams
  • White
  • Available clean processed and gamma sterilized or bulk packaged

Order Info

Clean and sterile covers are individually packaged in opaque bag and placed in double transparent poly liners in and ISO Class 4 cleanroom. Bulk covers are bulk packaged in double transparent poly liners.

Specifications

Size Medium
Material Tyvek
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Skid-resistant
Seam Type Serged
For Use With (Application) Barrier For Dry Particles, Non-hazardous Liquids And Microorganisms
Length (English) 10.75 in.
Length (Metric) 27.3 cm
Height (English) 18 in.
Height (Metric) 45.7 cm
Resistance Dry Particulates
Product Type Controlled Environments Boot Covers
Show More Show Less