missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks
GREENER_CHOICE

Catalog No. 17988081
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17988081 Medium
17988084 2X-Large
17988083 X-Large
17988082 Large
17988080 Small
17987100 3X-Large
17987101 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17988081 Supplier DuPont™ Supplier No. IC264SWHMD00300B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Should not be worn around open flame, heat source, sparks or ignition sources.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bulk packaged in double transparent poly liners.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette

Specifications

Size Medium
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Collar Type Bound
Size (English) Chest 32 to 36 in.
Size (Metric) Chest 81.28 to 91.44 cm
Sleeve Style Raglan Sleeve
Packaging Type Bulk Packaged
Length (Metric) 111.12 cm
Length (English) Sleeve 36.5 in.
Length (Metric) Sleeve 92.71 cm
Cleanroom Class Class 1000 (ISO 6)
Length (English) 43.75 in.
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less